Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem8a
Entrez Gene ID
303004
Gene Name
transmembrane protein 8A
Gene Aliases
M83, Tmem8
Chromosome Location
Chr.10: 15484951 - 15495380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106991.1 NP_001100461.1 9 - 10 1715 97
XM_006245973.2 XP_006246035.1 9 - 10 2329 97
XM_008767577.1 XP_008765799.1 9 - 10 3407 97

View Full Product Details