Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dach2
Entrez Gene ID
302333
Gene Name
dachshund family transcription factor 2
Gene Aliases
RGD1561687
Chromosome Location
Chr.X: 85086864 - 85484150 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257198.2 XP_006257260.1 12 - 13 2084 96

View Full Product Details