Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh19
Entrez Gene ID
317183
Gene Name
protocadherin 19
Gene Aliases
RGD1565392
Chromosome Location
Chr.X: 104387346 - 104493914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169129.1 NP_001162600.1 2 - 3 2473 96
XM_006257222.2 XP_006257284.1 3 - 4 2814 96
XM_006257223.2 XP_006257285.1 3 - 4 2814 96
XM_006257224.2 XP_006257286.1 2 - 3 2673 96

View Full Product Details