Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc3c1
Entrez Gene ID
317224
Gene Name
fibronectin type III domain containing 3C1
Gene Aliases
RGD1563846
Chromosome Location
Chr.X: 77416959 - 77559348 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191722.1 NP_001178651.1 - 3627 83

View Full Product Details