Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvrl2
Entrez Gene ID
308417
Gene Name
poliovirus receptor-related 2
Gene Aliases
-
Chromosome Location
Chr.1: 80631449 - 80666741 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012064.1 NP_001012064.1 6 - 7 1381 105
GenBank mRNA BC090075.1 - 6 - 7 1381 105

View Full Product Details