Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npas1
Entrez Gene ID
308387
Gene Name
neuronal PAS domain protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 78420646 - 78441841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107479.1 NP_001100949.1 7 - 8 1030 74
XM_006228413.2 XP_006228475.1 8 - 9 1158 74

View Full Product Details