Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bad
Entrez Gene ID
64639
Gene Name
BCL2-associated agonist of cell death
Gene Aliases
-
Chromosome Location
Chr.1: 222197829 - 222207454 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022698.1 NP_073189.1 3 - 4 495 75
XM_006230898.2 XP_006230960.1 3 - 4 393 75
GenBank mRNA AF031227.1 - 3 - 4 533 75
AF279910.1 - 3 - 4 495 75

View Full Product Details