The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108482.1 | NP_001101952.1 | 4 - 5 | 501 | 142 |
XM_006229098.2 | XP_006229160.1 | 4 - 5 | 1159 | 142 | |
XM_008759396.1 | XP_008757618.1 | 5 - 6 | 589 | 142 | |
XM_008759397.1 | XP_008757619.1 | 4 - 5 | 450 | 142 |