Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srrd
Entrez Gene ID
288717
Gene Name
SRR1 domain containing
Gene Aliases
RGD1562579
Chromosome Location
Chr.12: 50315863 - 50321488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_213799.8 XP_213799.6 5 - 6 738 101

View Full Product Details