Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcchc8
Entrez Gene ID
288661
Gene Name
zinc finger, CCHC domain containing 8
Gene Aliases
-
Chromosome Location
Chr.12: 38303662 - 38327174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105929.1 NP_001099399.1 5 - 6 591 100

View Full Product Details