Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irs2
Entrez Gene ID
29376
Gene Name
insulin receptor substrate 2
Gene Aliases
4PS, IRS-2
Chromosome Location
Chr.16: 83824515 - 83848569 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168633.1 NP_001162104.1 1 - 1 3323 80

View Full Product Details