Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lonrf1
Entrez Gene ID
306505
Gene Name
LON peptidase N-terminal domain and ring finger 1
Gene Aliases
RGD1562583
Chromosome Location
Chr.16: 59536248 - 59594251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771313.1 XP_008769535.1 7 - 8 1717 103

View Full Product Details