Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sele
Entrez Gene ID
25544
Gene Name
selectin E
Gene Aliases
-
Chromosome Location
Chr.13: 82355229 - 82365323 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138879.1 NP_620234.1 2 - 3 420 77
XM_008769608.1 XP_008767830.1 3 - 4 802 77
GenBank mRNA L25527.1 - 2 - 3 420 77
X69981.1 - 2 - 3 472 77

View Full Product Details