Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr76
Entrez Gene ID
311361
Gene Name
WD repeat domain 76
Gene Aliases
RGD1309616
Chromosome Location
Chr.3: 113445498 - 113479483 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234874.2 XP_006234936.1 6 - 7 1341 82
XM_006234875.2 XP_006234937.1 5 - 6 1248 82
XM_006234876.2 XP_006234938.1 5 - 6 786 82
XM_006234877.1 XP_006234939.1 5 - 6 779 82
XM_008762207.1 XP_008760429.1 2 - 3 458 82
XM_008762208.1 XP_008760430.1 3 - 4 380 82
XM_008762209.1 XP_008760431.1 2 - 3 207 82
XM_008762210.1 XP_008760432.1 2 - 3 145 82
XM_230512.7 XP_230512.4 5 - 6 817 82

View Full Product Details