Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akr1c12
Entrez Gene ID
361266
Gene Name
aldo-keto reductase family 1, member C12
Gene Aliases
-
Chromosome Location
Chr.17: 69614486 - 69647772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170342.1 NP_001163813.1 6 - 7 718 108
XM_006254190.2 XP_006254252.1 8 - 9 1568 108
XM_006254191.1 XP_006254253.1 9 - 10 1074 108
XM_008771873.1 XP_008770095.1 8 - 9 958 108
XM_008771874.1 XP_008770096.1 8 - 9 884 108
GenBank mRNA CN542908.1 - 3 - 4 108

Gene Symbol
Akr1c12l1
Entrez Gene ID
498790
Gene Name
aldo-keto reductase family 1, member C12-like 1
Gene Aliases
Akr1c17, RGD1559604
Chromosome Location
Chr.17: 69588119 - 69599168 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135744.1 NP_001129216.1 6 - 7 706 108
GenBank mRNA BC129122.1 - 6 - 7 706 108

View Full Product Details