The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134882.1 | NP_001128354.1 | 2 - 3 | 284 | 95 |
XM_006254276.2 | XP_006254338.1 | 2 - 3 | 401 | 95 | |
XM_006254278.2 | XP_006254340.1 | 2 - 3 | 204 | 95 | |
XM_008771888.1 | XP_008770110.1 | 2 - 3 | 187 | 95 |