Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gzmc
Entrez Gene ID
171290
Gene Name
granzyme C
Gene Aliases
Rnpk-4, Rnpk4
Chromosome Location
Chr.15: 35392158 - 35394787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134332.2 NP_599159.1 3 - 4 349 77
GenBank mRNA FQ219852.1 - 3 - 4 412 77
FQ219889.1 - 3 - 4 412 77
FQ219947.1 - 3 - 4 410 77
FQ219966.1 - 3 - 4 410 77
FQ219998.1 - 3 - 4 414 77
FQ220005.1 - 3 - 4 412 77
FQ220014.1 - 3 - 4 412 77
FQ220035.1 - 3 - 4 412 77
FQ220062.1 - 3 - 4 414 77
FQ220064.1 - 3 - 4 412 77
FQ220079.1 - 3 - 4 410 77
FQ220128.1 - 3 - 4 409 77
FQ220142.1 - 3 - 4 412 77
FQ220223.1 - 3 - 4 412 77
FQ220228.1 - 3 - 4 402 77
FQ220350.1 - 3 - 4 412 77
FQ220426.1 - 3 - 4 412 77
FQ220453.1 - 3 - 4 412 77
FQ220464.1 - 3 - 4 412 77
FQ220480.1 - 3 - 4 414 77
FQ220512.1 - 3 - 4 412 77
FQ220556.1 - 3 - 4 412 77
FQ220562.1 - 3 - 4 412 77
FQ220600.1 - 3 - 4 412 77
FQ220618.1 - 3 - 4 412 77
FQ220648.1 - 3 - 4 412 77
FQ220735.1 - 3 - 4 412 77
FQ220749.1 - 3 - 4 411 77
FQ220777.1 - 3 - 4 412 77
FQ220856.1 - 3 - 4 411 77
FQ220878.1 - 3 - 4 411 77
FQ220928.1 - 3 - 4 414 77
FQ221390.1 - 3 - 4 412 77
FQ221473.1 - 3 - 4 412 77
FQ221896.1 - 1 - 2 135 77
FQ221940.1 - 3 - 4 412 77
FQ223110.1 - 3 - 4 412 77
FQ223477.1 - 3 - 4 411 77
FQ228922.1 - 3 - 4 414 77
FQ229160.1 - 3 - 4 412 77
FQ229161.1 - 3 - 4 412 77
FQ229162.1 - 3 - 4 412 77
FQ229181.1 - 3 - 4 414 77
FQ229215.1 - 3 - 4 412 77
FQ229235.1 - 3 - 4 416 77
FQ229289.1 - 3 - 4 412 77
FQ229298.1 - 3 - 4 412 77
FQ229311.1 - 3 - 4 412 77
FQ229314.1 - 3 - 4 412 77
FQ229333.1 - 3 - 4 412 77
FQ229395.1 - 3 - 4 412 77
FQ229439.1 - 3 - 4 410 77
FQ229447.1 - 3 - 4 410 77
FQ229454.1 - 3 - 4 412 77
FQ229468.1 - 3 - 4 412 77
FQ229486.1 - 3 - 4 412 77
FQ229509.1 - 3 - 4 412 77
FQ229543.1 - 3 - 4 412 77
FQ229636.1 - 3 - 4 412 77
FQ229685.1 - 3 - 4 412 77
FQ229690.1 - 3 - 4 456 77
FQ229764.1 - 3 - 4 413 77
FQ229772.1 - 3 - 4 410 77
FQ229825.1 - 3 - 4 412 77
FQ229868.1 - 3 - 4 443 77
FQ229912.1 - 3 - 4 412 77
FQ229913.1 - 3 - 4 410 77
FQ230028.1 - 3 - 4 414 77
FQ230087.1 - 3 - 4 412 77
FQ230157.1 - 3 - 4 414 77
FQ230165.1 - 3 - 4 412 77
U57062.1 - 3 - 4 350 77

Gene Symbol
LOC502003
Entrez Gene ID
502003
Gene Name
similar to granzyme C
Gene Aliases
-
Chromosome Location
Chr.15: 35317747 - 35318247 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770722.1 XP_008768944.1 1 - 2 143 77

Gene Symbol
LOC691695
Entrez Gene ID
691695
Gene Name
similar to granzyme C
Gene Aliases
-
Chromosome Location
Chr.15: 36448970 - 36451720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751488.3 XP_003751536.2 3 - 4 470 77
GenBank mRNA FQ219891.1 - 3 - 4 412 77
FQ229899.1 - 3 - 4 412 77

View Full Product Details