Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myh6
Entrez Gene ID
29556
Gene Name
myosin, heavy chain 6, cardiac muscle, alpha
Gene Aliases
Myhca
Chromosome Location
Chr.15: 33605653 - 33629592 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017239.2 NP_058935.2 37 - 38 5840 87
GenBank mRNA BC070964.1 - - 1417 87
X15938.1 - 37 - 38 5837 87

View Full Product Details