The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108541.1 | NP_001102011.1 | 12 - 13 | 1169 | 62 |
XM_006231815.2 | XP_006231877.1 | 13 - 14 | 1415 | 62 | |
GenBank mRNA | BC166792.1 | - | 13 - 14 | 1341 | 62 |