Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcrla
Entrez Gene ID
304965
Gene Name
Fc receptor-like A
Gene Aliases
Fcrlm1, RGD1306176
Chromosome Location
Chr.13: 89296570 - 89306219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100682.1 NP_001094152.1 2 - 3 285 75
GenBank mRNA BC105902.1 - 2 - 3 256 75
BC158820.1 - 2 - 3 285 75
FQ225304.1 - 2 - 3 272 75
FQ227783.1 - 2 - 3 272 75
FQ232269.1 - 2 - 3 272 75
FQ233227.1 - 2 - 3 271 75

View Full Product Details