Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pyroxd1
Entrez Gene ID
297708
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
-
Chromosome Location
Chr.4: 176565733 - 176581971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004234.1 NP_001004234.1 4 - 5 444 118
GenBank mRNA BC079377.1 - 4 - 5 444 118

View Full Product Details