Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc34
Entrez Gene ID
499589
Gene Name
leucine rich repeat containing 34
Gene Aliases
RGD1565052
Chromosome Location
Chr.2: 116370047 - 116391964 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044696.1 NP_001038161.1 10 - 11 1323 127
XM_006232235.2 XP_006232297.1 11 - 12 1294 127
XM_008760937.1 XP_008759159.1 11 - 12 1385 127
XM_008760938.1 XP_008759160.1 10 - 11 1081 127
GenBank mRNA BC097432.1 - 10 - 11 1323 127

View Full Product Details