Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Depdc1b
Entrez Gene ID
310074
Gene Name
DEP domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.2: 39980802 - 40069982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107651.1 NP_001101121.1 1 - 2 131 78

View Full Product Details