Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
100359662
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.2: 27030295 - 27296860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760686.1 XP_008758908.1 14 - 15 1679 114

View Full Product Details