Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryba1
Entrez Gene ID
25583
Gene Name
crystallin, beta A1
Gene Aliases
BA3A1C, beta-A3
Chromosome Location
Chr.10: 65161152 - 65167504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013056.1 NP_037188.1 4 - 5 357 119
GenBank mRNA X15143.1 - 2 - 3 238 119

View Full Product Details