Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wsb1
Entrez Gene ID
303336
Gene Name
WD repeat and SOCS box-containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 66586905 - 66602984 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042561.1 NP_001036026.1 5 - 6 808 87
GenBank mRNA BC167746.1 - 5 - 6 811 87
CK473503.1 - 3 - 4 291 87

View Full Product Details