Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc52
Entrez Gene ID
289199
Gene Name
leucine rich repeat containing 52
Gene Aliases
RGD1312014
Chromosome Location
Chr.13: 85672013 - 85695421 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077434.1 NP_001070902.1 1 - 2 970 108
GenBank mRNA BC126099.1 - 1 - 2 970 108

View Full Product Details