Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab10
Entrez Gene ID
50993
Gene Name
RAB10, member RAS oncogene family
Gene Aliases
Ac1075
Chromosome Location
Chr.6: 27668387 - 27721120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017359.2 NP_059055.2 4 - 5 465 122
GenBank mRNA AY310140.1 - 2 - 3 229 122
AY724493.1 - 4 - 5 644 122
BC085744.1 - 4 - 5 465 122

View Full Product Details