The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031573.1 | NP_113761.1 | 8 - 9 | 871 | 105 |
GenBank mRNA | X07320.1 | - | 8 - 9 | 871 | 105 |