Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab3ip
Entrez Gene ID
29885
Gene Name
RAB3A interacting protein
Gene Aliases
RABIN3
Chromosome Location
Chr.7: 59927486 - 59971516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017313.3 NP_059009.2 6 - 7 1050 137
XM_006241357.2 XP_006241419.1 7 - 8 1219 137
XM_006241358.2 XP_006241420.1 7 - 8 1224 137
XM_006241359.2 XP_006241421.1 7 - 8 2228 137
XM_006241360.2 XP_006241422.1 7 - 8 1214 137
XM_008765382.1 XP_008763604.1 5 - 6 1180 137
XM_008765383.1 XP_008763605.1 6 - 7 659 137
GenBank mRNA BC127500.1 - 6 - 7 1053 137
U19181.1 - 6 - 7 1043 137

View Full Product Details