Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tsta3
- Entrez Gene ID
-
300036
- Gene Name
- tissue specific transplantation antigen P35B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 116958427 - 116963281 on Build Rnor_6.0
- Gene Symbol
- Pibf1
- Entrez Gene ID
-
306104
- Gene Name
- progesterone immunomodulatory binding factor 1
- Gene Aliases
-
RGD1305077
- Chromosome Location
-
Chr.15: 83494146 - 83663202 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ210517.1 |
- |
- |
|
70 |
View Full Product Details