Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsta3
Entrez Gene ID
300036
Gene Name
tissue specific transplantation antigen P35B
Gene Aliases
-
Chromosome Location
Chr.7: 116958427 - 116963281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127455.2 NP_001120927.1 9 - 10 852 72
NM_001270789.1 NP_001257718.1 9 - 10 918 72
NM_001270790.1 NP_001257719.1 8 - 9 789 72
GenBank mRNA BC158885.1 - 8 - 9 761 72
FQ210097.1 - 9 - 10 925 72
FQ221078.1 - 9 - 10 841 72

Gene Symbol
Pibf1
Entrez Gene ID
306104
Gene Name
progesterone immunomodulatory binding factor 1
Gene Aliases
RGD1305077
Chromosome Location
Chr.15: 83494146 - 83663202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ210517.1 - - 72

View Full Product Details