The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133543.3 | NP_598227.3 | - | 822 | 114 |
GenBank mRNA | BC088090.1 | - | - | 822 | 114 |
BC098650.1 | - | - | 884 | 114 | |
FQ209576.1 | - | - | 806 | 114 | |
FQ209710.1 | - | - | 841 | 114 | |
U18762.1 | - | - | 114 | ||
U33501.1 | - | - | 803 | 114 |