The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011933.1 | NP_001011933.1 | 9 - 10 | 1189 | 69 |
XM_006230020.2 | XP_006230082.1 | 10 - 11 | 1273 | 69 | |
GenBank mRNA | BC082015.1 | - | 9 - 10 | 1189 | 69 |