The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133380.2 | NP_596871.2 | 9 - 10 | 1019 | 66 |
XM_006230215.2 | XP_006230277.1 | 10 - 11 | 1170 | 66 | |
XM_006230216.2 | XP_006230278.1 | 8 - 9 | 960 | 66 | |
XM_008759862.1 | XP_008758084.1 | 9 - 10 | 6378 | 66 | |
XM_008759863.1 | XP_008758085.1 | 10 - 11 | 6325 | 66 | |
XM_008759864.1 | XP_008758086.1 | 10 - 11 | 6325 | 66 | |
GenBank mRNA | AB015746.1 | - | 10 - 11 | 1101 | 66 |
AB015747.1 | - | 9 - 10 | 1026 | 66 |