Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il4r
Entrez Gene ID
25084
Gene Name
interleukin 4 receptor
Gene Aliases
Il4ra
Chromosome Location
Chr.1: 196942345 - 196967221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB015746.1 - 7 - 8 841 68

View Full Product Details