Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubfd1
Entrez Gene ID
293454
Gene Name
ubiquitin family domain containing 1
Gene Aliases
RGD1306614, Ubph
Chromosome Location
Chr.1: 192025437 - 192040506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034911.1 NP_001030083.1 2 - 3 361 124
XM_006230189.2 XP_006230251.1 2 - 3 634 124
GenBank mRNA BC101866.1 - 2 - 3 361 124

View Full Product Details