Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp8
Entrez Gene ID
29172
Gene Name
aquaporin 8
Gene Aliases
AQP-8
Chromosome Location
Chr.1: 193424818 - 193430668 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019158.2 NP_062031.1 5 - 6 866 62
XM_006230187.2 XP_006230249.1 5 - 6 913 62
GenBank mRNA AB005547.1 - 5 - 6 865 62
AF007775.1 - 5 - 6 845 62
BC081812.1 - 5 - 6 866 62

View Full Product Details