Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gdpd3
Entrez Gene ID
293490
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
RGD1308386
Chromosome Location
Chr.1: 198199033 - 198210050 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728806.4 XP_002728852.3 2 - 3 771 152

View Full Product Details