Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcmt1
Entrez Gene ID
361643
Gene Name
leucine carboxyl methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.1: 193335633 - 193391620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199405.2 NP_955437.1 3 - 4 430 134
XM_006230191.2 XP_006230253.1 3 - 4 466 134
GenBank mRNA BC063186.1 - 3 - 4 433 134

View Full Product Details