Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrn4
Entrez Gene ID
311443
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
RGD1305256
Chromosome Location
Chr.3: 125521879 - 125547630 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_230550.8 XP_230550.4 3 - 4 2406 72

View Full Product Details