Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dfnb59
Entrez Gene ID
679552
Gene Name
deafness, autosomal recessive 59
Gene Aliases
-
Chromosome Location
Chr.3: 63508786 - 63518163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761977.1 XP_008760199.1 3 - 4 883 86

View Full Product Details