Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Avp
- Entrez Gene ID
-
24221
- Gene Name
- arginine vasopressin
- Gene Aliases
-
DI, VP, Vas
- Chromosome Location
-
Chr.3: 123117482 - 123119460 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M64785.1 |
- |
- |
254 |
99 |
View Full Product Details