Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gabpb1l
- Entrez Gene ID
-
364738
- Gene Name
- GA binding protein transcription factor, beta subunit 1-like
- Gene Aliases
-
Gabpb1, Gabpb2, Nrf-2beta, Nrf2b
- Chromosome Location
-
Chr.17: 52938851 - 52941213 on Build Rnor_6.0
- Gene Symbol
- Gabpb1
- Entrez Gene ID
-
499883
- Gene Name
- GA binding protein transcription factor, beta subunit 1
- Gene Aliases
-
RGD1560391
- Chromosome Location
-
Chr.3: 119091739 - 119135391 on Build Rnor_6.0
View Full Product Details