Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdhx
Entrez Gene ID
311254
Gene Name
pyruvate dehydrogenase complex, component X
Gene Aliases
Ac1164
Chromosome Location
Chr.3: 92910300 - 92933725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ232219.1 - - 267 83

View Full Product Details