The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100539.1 | NP_001094009.1 | 6 - 7 | 668 | 81 |
GenBank mRNA | BC158620.1 | - | 6 - 7 | 668 | 81 |
DQ403001.1 | - | 5 - 6 | 573 | 81 | |
FQ215046.1 | - | 6 - 7 | 664 | 81 |