The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277250.1 | NP_001264179.1 | 6 - 7 | 527 | 66 |
NM_001277251.1 | NP_001264180.1 | 7 - 8 | 586 | 66 | |
NM_001277252.1 | NP_001264181.1 | 6 - 7 | 467 | 66 | |
NM_001277253.1 | NP_001264182.1 | 6 - 7 | 579 | 66 | |
NM_057143.2 | NP_476484.1 | 6 - 7 | 709 | 66 | |
GenBank mRNA | AF157511.1 | - | 6 - 7 | 709 | 66 |
AF157512.1 | - | 6 - 7 | 463 | 66 | |
AJ007291.1 | - | 7 - 8 | 634 | 66 | |
FM055257.1 | - | 6 - 7 | 497 | 66 | |
FM061417.1 | - | 6 - 7 | 448 | 66 | |
FM069664.1 | - | 6 - 7 | 518 | 66 | |
FQ217993.1 | - | 2 - 3 | 156 | 66 | |
FQ220895.1 | - | 6 - 7 | 579 | 66 | |
FQ223564.1 | - | 6 - 7 | 577 | 66 | |
FQ224480.1 | - | 7 - 8 | 570 | 66 |