Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syndig1l
Entrez Gene ID
685882
Gene Name
synapse differentiation inducing 1-like
Gene Aliases
Tmem90a
Chromosome Location
Chr.6: 108392837 - 108415096 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109485.1 NP_001102955.1 1 - 2 284 74

View Full Product Details