The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006240356.2 | XP_006240418.1 | 4 - 5 | 683 | 71 |
XM_006240358.2 | XP_006240420.1 | 4 - 5 | 509 | 71 | |
XM_008764889.1 | XP_008763111.1 | 4 - 5 | 506 | 71 | |
XM_008764890.1 | XP_008763112.1 | 4 - 5 | 499 | 71 | |
XM_343089.7 | XP_343090.4 | 4 - 5 | 495 | 71 |