The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011977.1 | NP_001011977.1 | 10 - 11 | 1229 | 70 |
XM_006238860.1 | XP_006238922.1 | 10 - 11 | 1401 | 70 | |
XM_006238861.2 | XP_006238923.1 | 9 - 10 | 1162 | 70 | |
GenBank mRNA | BC078990.1 | - | 10 - 11 | 1229 | 70 |